missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPHK2 Recombinant Human Protein
SPHK2, along with SPHK1, catalyzes the phosphorylation of sphingosine producing the signalling molecule sphingosine-1-phosphate (S1P).
18240.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_064511.2 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 96.7kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428703
|
Invitrogen™
PR8935A |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15438783
|
Invitrogen™
PV5217 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapuetics for the treatment of cancer.
Specifications
NP_064511.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SPHK and DGCK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
96.7kDa | |
RUO - research use only | |
SPHK2 | |
Human | |
GST Tag | |
Frozen |