missing translation for 'onlineSavingsMsg'
Learn More

SPI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55100-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16876545

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to SPI1.

This gene encodes an ETS-domain transcription factor that activates gene expression during myeloid and B-lymphoid cell development. The nuclear protein binds to a purine-rich sequence known as the PU-box found near the promoters of target genes, and regulates their expression in coordination with other transcription factors and cofactors. The protein can also regulate alternative splicing of target genes. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

SPI1
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P17433, P17947, Q6BDS1
31 kDa transforming protein, OF, PU.1, SFPI1, SPI 1, SPI A, SPI1, Transcription factor PU.1
Rabbit
IgG
20 μL
-20°C
Primary
20375, 366126, 6688
Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
SPI1
Liquid
Spi1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers