Beskrivning
The SPINDOC Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifikationer
Specifikationer
| Celllinje | The lysate was created in HEK293T cells, using plasmid ID RC28181 and based on accession number NM_138471. The protein contains a C-terminal DDK tag. |
| För användning med (applikation) | Western Blot |
| Kvantitet | 0.1 mg |
| Förvaringsbuffert | RIPA buffer |
| Renhet | Protein |
| Testspecificitet | Homo sapiens chromosome 11 open reading frame 84 (C11orf84), mRNA. |
| Innehåll och lagring | -80°C. Avoid freeze-thaw cycles. |
| Proteintillstånd | Native |