Learn More
SPINK13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557668
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FNKRDFTRWP KPRCKMYIPL DPDYNADCPN VTAPVCASNG HTFQNECFFC VEQREFHYRI Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 53%.
SPINK13 Polyclonal antibody specifically detects SPINK13 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SPINK13 | |
Polyclonal | |
Unconjugated | |
SPINK13 | |
HBVDNAPTP1; Hepatitis B Virus (HBV) DNA polymerase transactivated protein 1; hepatitis B virus DNA polymerase transactivated human serine protease inhibitor; hepatitis B virus DNA polymerase transactivated serine protease inhibitor; Hespintor; LiESP6; serine peptidase inhibitor, Kazal type 13 (putative); serine protease inhibitor Kazal-type 13; Serine protease inhibitor Kazal-type 5-like 3; SPINK13; SPINK5L3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
153218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q1W4C9 | |
SPINK13 | |
Recombinant protein corresponding to Human SPINK13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only