Learn More
SPINK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555426
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KYRTPNCSQY RLPGCPRHFN PVCGSDMSTY ANECTLCMKI REGGHNIKII RNGPC Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 58%.
SPINK2 Polyclonal antibody specifically detects SPINK2 in Human samples. It is validated for Immunohistochemistry.Specifications
SPINK2 | |
Polyclonal | |
Unconjugated | |
SPINK2 | |
1700007F22Rik; acrosin-trypsin inhibitor; AV038945; Epididymis tissue protein Li 172; HUSI-II; serine peptidase inhibitor, Kazal type 2; serine peptidase inhibitor, Kazal type 2 (acrosin-trypsin inhibitor); serine protease inhibitor Kazal-type 2; SPINK2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6691 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P20155 | |
SPINK2 | |
Recombinant protein corresponding to Human SPINK2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only