Learn More
Buerkle™ Spiral Screw for PowderProof™ Sampler
Buerkle™ Spiral Screw for PowderProof™ Sampler is a spare part for use with Buerkle™ PowderProof™ Sampler.
Brand: Buerkle™ 5302-0611
Additional Details : Weight : 0.22700kg
Description
- Spare part for Buerkle™ PowderProof™ Sampler
Specifications
Spiral Screw | |
Stainless Steel (V4A) | |
PowderProof for Granules From 2 to 4 mm dia | |
60 cm |