Läs mer
Spirapril Hydrochloride, TRC
Produktkod. 30427176
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427176 10 mg
30411832 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30427176 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68250010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 94841-17-5
Molekylformel C22 H30 N2 O5 S2 . Cl H
Molekylvikt (g/mol) 503.07
Synonym (8S)-7-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,4-dithia-7-azaspiro[4.4]nonane-8-carboxylic acid hydrochloride (1:1), [8S-[7[R*(R*)],8R*]]-7-[2-[[1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,4-dithia-7-azaspiro[4.4]nonane-8-carboxylic acid monohydrochloride, Renormax, Renpress, Sandopril, Spirapril hydrochloride, TI 211-950
IUPAC-namn (8S)-7-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-1,4-dithia-7-azaspiro[4.4]nonane-8-carboxylic acid;hydrochloride
LEDER Cl.CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2CC3(C[C@H]2C(=O)O)SCCS3
Specifikationer
Kemiskt namn eller material Spirapril hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C22H30N2O5S2.ClH/c1-3-29-21(28)17(10-9-16-7-5-4-6-8-16)23-15(2)19(25)24-14-22(30-11-12-31-22)13-18(24)20(26)27;/h4-8,15,17-18,23H,3,9-14H2,1-2H3,(H,26,27);1H/t15-,17-,18-;/m0./s1
Formel vikt 502.1363