Learn More
SPIRE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559274
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LEEIKAERKL RPVSPEEIRR SRLDVTTPES TKNLVESSMV NGGLTSQTKE NGLSTSQQVP AQRKKLLRAP TLAELDSSES EEETLHK Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.
SPIRE1 Polyclonal antibody specifically detects SPIRE1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SPIRE1 | |
Polyclonal | |
Unconjugated | |
SPIRE1 | |
KIAA1135; Protein spire homolog 1; SPIR1; Spir-1; spire actin nucleation factor 1; spire family actin nucleation factor 1; spire homolog 1; spire type actin nucleation factor 1; SPIRE1; spire-type actin nucleation factor 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56907 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q08AE8 | |
SPIRE1 | |
Recombinant protein corresponding to Human SPIRE1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only