Läs mer
Spiro[androsta-1,4-diene-6,2'-oxirane]-3,17-dione(Mixture of Diastereomers), TRC
Produktkod. 30136322
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136322 25 mg
30134687 50 mg
30004921 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136322 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68296025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 184972-12-1
Molekylformel C20 H24 O3
Molekylvikt (g/mol) 312.4
Synonym Spiro[androsta-1,4-diene-6,2'-oxirane]-3,17-dione, 6-Spirooxiran-androsta-1,4-diene-3,17-dione
IUPAC-namn (8R,9S,10R,13S,14S)-10,13-dimethylspiro[7,8,9,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-6,2'-oxirane]-3,17-dione
LEDER C[C@]12CC[C@H]3[C@@H](CC4(CO4)C5=CC(=O)C=C[C@]35C)[C@@H]1CCC2=O
Specifikationer
Kemiskt namn eller material Spiro[androsta-1,4-diene-6,2'-oxirane]-3,17-dione(mixture of diastereomers)
Kvantitet 25 mg
InChI formel InChI=1S/C20H24O3/c1-18-7-5-12(21)9-16(18)20(11-23-20)10-13-14-3-4-17(22)19(14,2)8-6-15(13)18/h5,7,9,13-15H,3-4,6,8,10-11H2,1-2H3/t13-,14-,15-,18+,19-,20?/m0/s1
Formel vikt 312.1725