missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPON1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 720248
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Monkey, Dog, Sheep, Bovine
SPON1 is a cell adhesion protein that promotes the attachment of spinal cord and sensory neuron cells and the out growth of neurites in vitro. SPON1 may contribute to the growth and guidance of axons in both the spinal cord and the PNS. Is known to be a major factor for vascular smooth muscle cells.Specifications
SPON1 | |
Polyclonal | |
Unconjugated | |
SPON1 | |
KIAA0762, VSGP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10418, 233744, 64456 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4 | |
Q9HCB6, Q8VCC9, P35446 | |
SPON1 | |
Peptides corresponding to human SPON1 (aa216-229, 102-120, 782-800) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |