missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPOP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SPOP |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836484
|
Proteintech
16750-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826484
|
Proteintech
16750-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.Specifications
SPOP | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43791, Q6ZWS8 | |
20747, 287643, 8405 | |
SPOP Fusion Protein Ag10215 | |
Primary | |
-20°C | |
SPOP |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HIB homolog 1, Roadkill homolog 1, speckle type POZ protein, SPOP, TEF2 | |
Spop | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |