missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPOP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16750-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.Specifications
SPOP | |
Polyclonal | |
Unconjugated | |
SPOP | |
HIB homolog 1, Roadkill homolog 1, speckle type POZ protein, SPOP, TEF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20747, 287643, 8405 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43791, Q6ZWS8 | |
Spop | |
SPOP Fusion Protein Ag10215 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |