Learn More
Uvex Sport Black PU 65904 Low Shoe - Width 14
Offers first-class wearer comfort with a sporty design; low weight, climate-optimizes high-tech materials and ergonomic outsole
1575.00 SEK
Specifications
Color | Black |
---|---|
Material | Textile |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17282541
|
Uvex
6590435 |
35 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17292541
|
Uvex
6590436 |
36 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17202551
|
Uvex
6590437 |
37 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17212551
|
Uvex
6590438 |
38 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17222551
|
Uvex
6590439 |
39 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17232551
|
Uvex
6590440 |
40 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17242551
|
Uvex
6590441 |
41 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17252551
|
Uvex
6590442 |
42 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17262551
|
Uvex
6590443 |
43 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17272551
|
Uvex
6590444 |
44 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17282551
|
Uvex
6590445 |
45 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17292551
|
Uvex
6590446 |
46 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17202561
|
Uvex
6590447 |
47 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17212561
|
Uvex
6590448 |
48 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17222561
|
Uvex
6590449 |
49 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17232561
|
Uvex
6590450 |
50 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17242561
|
Uvex
6590451 |
51 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
17252561
|
Uvex
6590452 |
52 |
1575.00 SEK
1 pair |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
Description
Uvex 1 Sport Shoe 6590.4 S1 P SRC
- Modern, ultra-lightweight and flexible S1 safety shoe
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials are free of silicones plasticizers and other paint wetting impairment substances
- Meets ESD specification with leakage resistance less than 35 megohms
- 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good lateral stability, no thermal conductivity
- Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
- Penetration-resistant, non-metallic midsole in accordance with latest safety standards, which does not restrict flexibility
- Made with new developed technology for exceptional wearer comfort
- Uvex Climazone technology for optimum wearer comfort
- Virtually seam-free construction made from high-tech materials to eliminate pressure points
- Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
- Soft, padding on collar and tongue
- For light applications
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6590.4
- Standard EN ISO 20345:2011 S1 P SRC
- Sizes: 35 to 52
Specifications
Black | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Textile | |
Composite | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S1 |