Learn More
Uvex Sport Hygiene White PU 65817 Low Shoe - Width 10
Modern, metal free, ultra-lightweight and flexible S2 safety slip-on shoe
1355.00 SEK
Specifications
Color | White |
---|---|
Material | Microfiber |
Toe Material | Composite |
Disposable | No |
Certifications/Compliance (Europe) | EN ISO 20345:2011 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17262381
|
Uvex
6581735 |
35 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17272381
|
Uvex
6581736 |
36 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17282381
|
Uvex
6581737 |
37 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17292381
|
Uvex
6581738 |
38 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17202391
|
Uvex
6581739 |
39 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17212391
|
Uvex
6581740 |
40 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17222391
|
Uvex
6581741 |
41 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17232391
|
Uvex
6581742 |
42 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17242391
|
Uvex
6581743 |
43 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17252391
|
Uvex
6581744 |
44 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17262391
|
Uvex
6581745 |
45 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17272391
|
Uvex
6581746 |
46 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17282391
|
Uvex
6581747 |
47 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17292391
|
Uvex
6581748 |
48 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17202401
|
Uvex
6581749 |
49 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17212401
|
Uvex
6581750 |
50 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17222401
|
Uvex
6581751 |
51 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17232401
|
Uvex
6581752 |
52 |
1355.00 SEK
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
Uvex 1 Sport Hygeine Shoe 6581.7 S2 SRC
- Upper made from water-repellent microfiber material that is easy to clean
- Minimum number of seams to reduce contamination
- Made from synthetic material, suitable for people allergic to chrome
- Washable up to 30°C
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Meets ESD specification with volume
- Resistance less than 35 megohms
- 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good lateral stability, no thermal conductivity
- Ergonomically designed outsole comprising dual-density polyurethane with good shock, absorption and excellent slip resistance
- Breathable materials
- Virtually seam-free construction to eliminate pressure points
- Removable anti-static comfort insole with moisture transport system and additional shock absorption around heel and forefoot
- Soft padding on collar and tongue
- For light applications
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6581.7
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 35 to 52
Specifications
White | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S2 |
Microfiber | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |