Learn More
Uvex Sport NC 95179 Insole - Width 12
Comfortable climatic insole with antistatic properties for Uvex 1 Sport NC
Brand: Uvex 9517939
Additional Details : Weight : 0.08700kg
Description
- Full-surface foot shock absorption
- Highly breathable and moisture-absorbent material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No: 9517.9
- Sizes: 35 to 52
Specifications
Black | |
Microvelour | |
No | |
Wide width | |
No |
Uvex 1 Sport NC | |
39 | |
Unisex | |
Yes |