Beskrivning
- Sporty, lightweight safety spectacles with contoured lens for a great fit on narrow or wider faces
- Robust polycarbonate lens with proven uvex supra-vision coating technology - permanently anti-fog and scratch-resistant lenses
- Combination of soft padding around the brow and ergonomically shaped side arms with soft ear-pieces ensures a comfortable, secure fit without creating pressure points
- Soft, adjustable nose piece allows spectacles to be adapted to the wearer to deliver a comfortable, non-slip and pressure-free fit
- Certified in accordance with EN 166 (personal eye protection) and EN 172 (sunglare filters for industrial use) UV400 - 100% protection against harmful UV radiation to 400 nm
Specifikationer
Specifikationer
| Linsbeläggning | Anti-fog, Scratch-resistant, Anti-scratch |
| Linsfärg | Silver, Mirror |
| Produkttyp | Safety Glasses |
| Material | Polycarbonate |
| Disponibel | Reusable |
| Linsmaterial | Polycarbonate |
| Rammaterial | TPE |
| UV-skydd | UV400 |
| Ramtyp | Semi-rimless |