Läs mer
Fisherbrand™ SPP Disposable Beard Cover
General purpose, high quality, non-woven disposable beard covers that are made of spunbound polypropylene material, ideal for use in the laboratory or elsewhere for promoting general hygiene.
Brand: Fisherbrand™ FBHMBC
712.45 SEK valid until 2026-08-31
BÄSTA PRIS på kampanj! Använd kampanjkod: "28036" för att få ditt kampanjpris.
Beskrivning
General purpose beard covers with integral elasticated headloop and seam for a comfortable, universal fit.
- Non-woven, lightweight spunbound polypropylene at 12 g/m2
- For use in the laboratory or wider working environment
- Manufactured and vacuum-sealed in a stringent clean environment
- Latex-free
- ISO tested to ISO3801 and ISO13934-1
Specifikationer
| Beard Cover | |
| Spunbound Polypropylene | |
| Yes | |
| Elastic | |
| General Purpose in the Lab or Cleanroom | |
| 23 ±2 cm; 46 ±2 cm Stretched | |
| 12 gsm |
| Large/Universal | |
| White | |
| Yes | |
| PPE CAT I, ISO Tested To ISO3801 and ISO13934-1; Meets 4.0 AQL Level II Sampling Plan Per MIL-std-150E For Defect Criteria | |
| 1000 Pk. | |
| 10 cm ±2 cm |