missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRY3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549294
Additional Details : Weight : 0.01000kg
Description
SPRY3 Polyclonal antibody specifically detects SPRY3 in Human samples. It is validated for Western Blot.Specifications
SPRY3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O43610 | |
SPRY3 | |
KLH conjugated synthetic peptide between 261-288 amino acids from the C-terminal region of human SPRY3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SPRY3 | |
antagonist of FGF signaling; Gm1409; Gm391; Protein sprouty homolog 3; RGD1562172; sprouty homolog 3; sprouty homolog 3 (Drosophila); sprouty RTK signaling antagonist 3; sprouty3; SPRY3; spry-3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10251 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |