Learn More
SPTLC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565493
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MANPGGGAVCN GKLHNHKKQS NGSQSRNCTK NGIVKEAQQN GKPHFYDKLI VESFEEA Highest antigen sequence identity to the following orthologs - mouse 53%, rat 52%.
SPTLC3 Polyclonal antibody specifically detects SPTLC3 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
SPTLC3 | |
Polyclonal | |
Unconjugated | |
SPTLC3 | |
C130053K05Rik; C20orf38; dJ718P11; dJ718P11.1; hLCB2b; LCB 3; LCB2b; LCB3; long chain base biosynthesis protein 2b; Long chain base biosynthesis protein 3; serine palmitoyltransferase 3; serine palmitoyltransferase long chain base subunit 3; serine palmitoyltransferase, long chain base subunit 2-like (aminotransferase 2); serine palmitoyltransferase, long chain base subunit 3; serine-palmitoyl-CoA transferase 3; Serine-palmityl-CoA transferase 3; SPT 3; SPT3; SPTLC2L; SPTLC3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55304 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NUV7 | |
SPTLC3 | |
Recombinant Human SPTLC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only