Läs mer
Tocris Bioscience™ SQ 22536
GREENER_CHOICE
Produktkod. 30001428
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001428 10 mg
18044582 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001428 Leverantör Tocris Bioscience™ Leverantörsnummer 1435/10
Adenylyl cyclase inhibitor
Inhibitor of adenylyl cyclase (IC50 = 1.4 μM). Inhibits PGE1-stimulated increases in cAMP levels in intact human platelets. Cell permeable.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17318-31-9
Molekylformel C9H11N5O
Specifikationer
Kemiskt namn eller material SQ 22536
Kvantitet 10 mg
Formel vikt 205.22
Renhet >99%
Molekylvikt (g/mol) 205.22