missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Squalene, >98%, MP Biomedicals™
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Specifications
Color | Colorless |
Density | 0.855g/mL |
Quantity | 10 mL |
Formula Weight | 410.7 |
Physical Form | Liquid |
Chemical Name or Material | Squalene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'