Läs mer
Tocris Bioscience™ SR 10067
Selective Rev-Erbα/β agonist; anxiolytic
Brand: Tocris Bioscience™ 6038/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SR 10067 is a selective Rev-Erbα/β agonist (IC50 values are 160 and 170 nM for Rev-Erbβ and Rev-Erbα respectively). Exhibits no significant activity against a wide range of nuclear receptors, other receptors, ion channels and transporters at 20 μM. Increases wakefulness, reduces REM and slow wave sleep in vivo. Also reduces anxiety-like behavior in marble-burying assay. Cell permeable and brain penetrant.Specifikationer
| [3-[[4-(1,1-Dimethylethoxy)phenoxy]methyl]-3,4-dihydro-2(1H)-isoquinolinyl]-1-naphthalenylmethanone | |
| 1380548-02-6 | |
| >98% | |
| 10 mg |