missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ SR 48692

Beskrivning
SR 48692 is a neurotensin antagonist; selective for NTS1 over NTS2 (apparent affinity, Ke, is 36 nM for NTS1). Competitively inhibits binding of [125I]-neurotensin to HT29 and N1E115 cell membranes (IC50 values are 15.3 and 20.4 nM respectively). Orally bioavailable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-[[[1-(7-Chloro-4-quinolinyl)-5-(2,6-dimethoxyphenyl)-1H-pyrazol-3-yl]carbonyl]amino]-tricyclo[3.3.1.13,7]decane-2-carboxylic acid |
| CAS | 146362-70-1 |
| Analysprocentintervall | >98% |
| Kvantitet | 25 mg |
| Molekylformel | C32H31ClN4O5 |
| Formel vikt | Observed MW: 587.07 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?