missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ SR 58611A hydrochloride
GREENER_CHOICE

Produktkod. 18095842
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18095842 50 mg
18053892 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18095842 Leverantör Tocris Bioscience™ Leverantörsnummer 3986/50

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

β3 agonist; displays anxiolytic and antidepressant effects

Selective β3-adrenergic receptor agonist. Displays both anxiolytic and antidepressant effects in rodent models. Orally active and brain penetrant.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 121524-09-2
Molekylformel C22H27Cl2NO4

Specifikationer

Kemiskt namn eller material SR 58611A hydrochloride
Kvantitet 50 mg
Formel vikt 440.36
Renhet >98%
Molekylvikt (g/mol) 440.361
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.