Läs mer
Tocris Bioscience™ SR 59230A hydrochloride
GREENER_CHOICE
Produktkod. 18064272
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18064272 50 mg
18044152 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18064272 Leverantör Tocris Bioscience™ Leverantörsnummer 1511/50
Potent and selective β3 antagonist
SR 59230A hydrochloride is a potent and selective β3 adrenoceptor antagonist (IC50 values are 40, 408 and 648 nM for β3, β1 and β2 receptors respectively). Orally active in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1135278-41-9
Molekylformel C21H28ClNO2
Specifikationer
Kemiskt namn eller material SR 59230A hydrochloride
Kvantitet 50 mg
Formel vikt 361.91
Renhet >98%
Molekylvikt (g/mol) 361.91