Läs mer
Tocris Bioscience™ SR 7826
Potent and selective LIMK inhibitor; antitumor
Brand: Tocris Bioscience™ 5626/50
This item is not returnable.
View return policy
Description
SR 7826 is a potent and selective LIMK inhibitor (IC50 values are 43, 5536 and 6565 nM for LIMK1, ROCKI and ROCKII, respectively). Inhibits cofilin phosphorylation in A7r5 cells and suppresses migration and invasion of PC-3 cells in vitro. Orally bioavailable.Specifications
| N-(2-Hydroxyethyl)-N′-[4,5-methyl-7H-pyrrolo[2,3-d]pyrimidine-4-yl)phenyl]-N-phenylurea | |
| 1219728-20-7 | |
| 50 mg |