Learn More
Tocris Bioscience™ SR 7826
Potent and selective LIMK inhibitor; antitumor
Brand:  Tocris Bioscience™ 5626/10
Additional Details : CAS Number : 1219728-20-7 Weight : 0.00970kg
 View more versions of this product
Product Code. 18700498
  • 3003.16 SEK / 10mg
Estimated Shipment: 31-07-2024
to see stock.
Description
Description
SR 7826 is a potent and selective LIMK inhibitor (IC50 values are 43, 5536 and 6565 nM for LIMK1, ROCKI and ROCKII, respectively). Inhibits cofilin phosphorylation in A7r5 cells and suppresses migration and invasion of PC-3 cells in vitro. Orally bioavailable.
Specifications
Specifications
N-(2-Hydroxyethyl)-N′-[4,5-methyl-7H-pyrrolo[2,3-d]pyrimidine-4-yl)phenyl]-N-phenylurea
10 mg
C22H21N5O2
>98%
1219728-20-7
LIMK Inhibitors
387.43