Learn More
SR9243, TRC
Product Code. 30425976
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425976 10 mg 10mg
30413038 50 mg -
30420410 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30425976 Supplier Toronto Research Chemicals Supplier No. TRCS68447510MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1613028-81-1
Molecular Formula C31H32BrNO4S2
Molecular Weight (g/mol) 626.62
Synonym SR9243, N-[2-(3-Bromophenyl)ethyl]-2,4,6-trimethyl-N-[[3'-(methylsulfonyl)[1,1'-biphenyl]-4-yl]methyl]benzenesulfonamide
IUPAC Name N-[2-(3-bromophenyl)ethyl]-2,4,6-trimethyl-N-[[4-(3-methylsulfonylphenyl)phenyl]methyl]benzenesulfonamide
SMILES Cc1cc(C)c(c(C)c1)S(=O)(=O)N(CCc2cccc(Br)c2)Cc3ccc(cc3)c4cccc(c4)S(=O)(=O)C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C31H32BrNO4S2/c1-22-17-23(2)31(24(3)18-22)39(36,37)33(16-15-25-7-5-9-29(32)19-25)21-26-11-13-27(14-12-26)28-8-6-10-30(20-28)38(4,34)35/h5-14,17-20H,15-16,21H2,1-4H3
Formula Weight 625.0956
Chemical Name or Material N-[2-(3-Bromophenyl)ethyl]-2,4,6-trimethyl-N-[(3'-(methylsulfonyl)[1,1'-biphenyl]-4-yl)methyl]benzenesulfonamide