Learn More
Invitrogen™ Human SRC, His Tag Recombinant Protein
SRC Recombinant Human Protein
5800.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_005408 |
---|---|
Gene Alias | SRC |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11539286
|
Invitrogen™
P3044 |
10μg |
5800.00 SEK
10µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488393
|
Invitrogen™
PR4336E |
100 μg |
18220.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10760355
|
Invitrogen™
PV3584 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SRC is a cytoplasmic tyrosine kinase and proto-oncogene involved in the regulation of embryonic development and cell growth.Specifications
NP_005408 | |
Human | |
Baculovirus |
SRC | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.