Learn More
SRC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566314
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: PKLERLDSKTD PASNTKLIAM KTEKEEMSFE PGDQPGSELD NLEEILDDLQ NSQLPQLFPD TRPGAPAGSV Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
SRC2 Polyclonal antibody specifically detects SRC2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
SRC2 | |
Polyclonal | |
Unconjugated | |
NCOA2 | |
bHLHe75; c-fgr; Class E basic helix-loop-helix protein 75; c-src2; D1Ertd433e; FLJ43153; Glucocorticoid receptor-interacting protein 1; glucocorticoid receptor-interacting protein-1; Grip1; GRIP-1; hTIF2; I79_019839; KAT13C; MGC75096; NCOA2; NCoA-2; Nuclear receptor coactivator 2; p160 steroid receptor coactivator 2; p55c-fgr; p58c-fgr; SRC2; SRC-2; Steroid receptor coactivator 2; TIF 2; Tif2; transcriptional intermediary factor 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10499 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15596 | |
NCOA2 | |
Recombinant Human SRC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only