missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
SREBP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-41282
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SREBP2 Polyclonal specifically detects SREBP2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifikationer
| SREBP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SREBF2 | |
| Antibody was raised against a 15 amino acid peptide near the center of human SREBF2. The immunogen is located within amino acids 440 - 490 of SREBF2. Amino Acid Squence: LDDAKVKDEPDSPPV | |
| Affinity Purified | |
| RUO | |
| 6721 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Immunofluorescence 20 μg/mL | |
| BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2 | |
| Rabbit | |
| 128 kDa | |
| 0.1 mg | |
| Primary | |
| At least three isoforms of SREBF2 are known to exist. SREBF2 antibody is predicted not to cross-react with SREBF1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering