Learn More
SREK1IP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561066
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MAVPGCNKDS VRAGCKKCGY PGHLTFECRN FLRVDPKRDI VLDVSSTSSE DS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
Possible splicing regulator involved in the control of cellular survival.Specifications
SREK1IP1 | |
Polyclonal | |
Unconjugated | |
SREK1IP1 | |
p18 splicing regulatory protein; P18SRP; protein SFRS12IP1; protein SREK1IP1; SFRS12-interacting protein 1; SFRS12IP1; Splicing regulatory protein of 18 kDa; SREK1 interacting protein 1; SREK1-interacting protein 1; SREK1IP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
285672 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N9Q2 | |
SREK1IP1 | |
Recombinant protein corresponding to Human SREK1IP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only