Läs mer
SRI 31215 TFA, MedChemExpress
Produktkod. 16491077
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16491077 50 mg -
16454818 10 mg 10mg
16447567 5 mg -
16435616 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16491077 Leverantör Medchem Express Leverantörsnummer HY114363A
MedChemExpress SRI 31215 (TFA), a triplex inhibitor of matriptase, hepsin and hepatocyte growth factor activator (HGFA) with IC50s of 0.69 μM, 0.65 μM, 0.3 μM, blocks pro-HGF activation and thus mimics the activity of HAI-1/2.
SRI 31215 (TFA), a triplex inhibitor of matriptase, hepsin and hepatocyte growth factor activator (HGFA) with IC50s of 0.69 μM, 0.65 μM, 0.3 μM, blocks pro-HGF activation and thus mimics the activity of HAI-1/2.
Specifikationer
Kemiskt namn eller material SRI 31215 TFA
CAS 1832686-44-8
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C27H34F3N5O3
Kvantitet 50 mg
Löslighetsinformation DMSO : 125 mg/mL (234.26 mM; Need ultrasonic)
LEDER N=C(C1=CC=CC(N2CC(C)CN(CC3CCN(CC4=CC=CC=C4)CC3)C2=O)=C1)N.OC(C(F)(F)F)=O
Molekylvikt (g/mol) 533.59
Formel vikt 533.59
Procent renhet 98.81%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation