Learn More
SRP72 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556994
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ENALKTIESA NQQTDKLKEL YGQVLYRLER YDECLAVYRD LVRNSQDDYD EERKTNLSAV VAAQSNWEKV VPENLGLQEG THELCYNTAC A Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
SRP72 Polyclonal antibody specifically detects SRP72 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SRP72 | |
Polyclonal | |
Unconjugated | |
SRP72 | |
5730576P14Rik; 72kDa; AI132477; BC019196; BMFF; BMFS1; C77589; epididymis luminal protein 103; HEL103; signal recognition particle 72; signal recognition particle 72 kDa; signal recognition particle 72 kDa protein; signal recognition particle 72kD; signal recognition particle 72kDa; signal recognition particle subunit SRP72; SRP72 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6731 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O76094 | |
SRP72 | |
Recombinant protein corresponding to Human SRP72 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only