missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRPK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14073-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a serine/arginine protein kinase specific for the SR (serine/arginine-rich domain) family of splicing factors. The protein localizes to the nucleus and the cytoplasm. It is thought to play a role in regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors. A second alternatively spliced transcript variant for this gene has been described, but its full length nature has not been determined.Specifications
SRPK1 | |
Polyclonal | |
Unconjugated | |
SRPK1 | |
SFRS protein kinase 1, SFRSK1, SR protein specific kinase 1, SRPK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20815, 361811, 6732 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O70551, Q96SB4 | |
Srpk1 | |
SRPK1 Fusion Protein Ag5149 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |