missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRSF7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563368
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EKGHYAYDCH RYSRRRRSRS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
SRSF7 Polyclonal antibody specifically detects SRSF7 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SRSF7 | |
Polyclonal | |
Unconjugated | |
Srsf7 | |
35kDa; 9430065L19Rik; 9G8; AAG3; aging-associated protein 3; hypothetical protein LOC507066; NX-96; serine and arginine rich splicing factor 7; serine/arginine-rich splicing factor 7; serine/arginine-rich splicing factor 7-like protein; SFRS7; Splicing factor 9G8; splicing factor, arginine/serine-rich 7; splicing factor, arginine/serine-rich 7, 35kDa; SR splicing factor 7; SRSF7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6432 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16629 | |
Srsf7 | |
Recombinant protein corresponding to Human SRSF7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only