Learn More
Paragon Scientific Sucrose Brix/RI Standards
Paragon ISO 17025 and ISO 17034 certified brix standards are for the calibration and verification of all types of refractometer e.g handheld, Abbe and high accuracy digital instruments. Typically used within the food and beverage and pharmaceutical industries.
Brand:  Paragon Scientific SS60
Additional Details : Weight : 0.20000kg
Concentration 60.00 Brix
 View more versions of this product
Product Code. 16060420
  • 507.00 SEK / 15mL
Estimated Shipment
16-02-2021
Add to basket
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Primary level refractive index standards traceable to National Standards
  • Manufactured in accordance with ICUMSA Methods
  • Supplied in tamper evident packs with 6 weeks shelf life
Specifications
Chemical Identifiers
15 mL
Specifications
60.00 Brix
Odorless
Colorless
Liquid
Sucrose
15 mL
Videos
SDS
Documents
Nominal values quoted