missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSPN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26685-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Component of the dystrophin-glycoprotein complex (DGC), a complex that spans the muscle plasma membrane and forms a link between the F-actin cytoskeleton and the extracellular matrix. Preferentially associates with the sarcoglycan subcomplex of the DGC.Specifications
SSPN | |
Polyclonal | |
Unconjugated | |
SSPN | |
DAGA5, Kirsten ras associated protein, KRAG, ospan, NSPN, Sarcospan, SPN1, SPN2, SSPN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16651, 500364, 8082 | |
Antibody | |
IgG |
Western Blot | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14714, Q62147 | |
Sspn | |
SSPN Fusion Protein Ag24511 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |