Läs mer
Tocris Bioscience™ SSR 180711 hydrochloride
Partial agonist at α7 nAChR
Brand: Tocris Bioscience™ 5559/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SSR 180711 hydrochloride is a partial agonist at α7 nAChR. Enhances episodic memory and reverses MK-801 -induced deficits in retention of episodic memory. Increases extracellular dopamine levels in rat prefrontal cortex. Also exhibits antidepressant-like properties.Specifikationer
| 4-Bromophenyl 1,4-diazabicyclo[3.2.2]nonane-4-carboxylate hydrochloride | |
| 446031-79-4 | |
| 10 mg |