missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSTR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SSTR1 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850145
|
Proteintech
20587-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840145
|
Proteintech
20587-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Somatostatin acts to regulate numerous physiological processes by binding to and activating specific receptors in target tissues. Activation of these receptors by somatostatin, which is secreted by nerve and endocrine cells, regulates the secretion of insulin, glucagon and growth hormone, neuronal excitability in both the brain and the peripheral nervous system, and cell growth. Somatostatin receptors have been implicated in numerous diseases ranging from Alzheimer′s to cancers of the gastrointestinal tract, breast, prostate, and pituitary.Specifications
SSTR1 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P28646, P30872, P30873 | |
20605, 25033, 6751 | |
SSTR1 Fusion Protein Ag14287 | |
Primary | |
-20°C | |
SSTR1 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
somatostatin receptor 1, Somatostatin receptor type 1, SRIF 2, SS 1 R, SS1 R, SS1R, SSTR1 | |
SSTR1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |