missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
SSX2 Polyclonal Antibody for ELISA, IHC (P), Western Blot
SSX2 belongs to the family of highly homologous synovial sarcoma X (SSX) breakpoint proteins. These proteins may function as transcriptional repressors. They are also capable of eliciting spontaneously humoral and cellular immune responses in cancer patients, and are potentially useful targets in cancer vaccine-based immunotherapy. SSX1, SSX2 and SSX4 genes have been involved in the t(X;18) translocation characteristically found in all synovial sarcomas. This translocation results in the fusion of the synovial sarcoma translocation gene on chromosome 18 to one of the SSX genes on chromosome X. The encoded hybrid proteins are probably responsible for transforming activity. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifikationer
Specifikationer
| Antigen | SSX2 |
| Användningsområden | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 0.32 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | Ssx2 |
| Genaccessionsnr. | Q16385 |
| Gene Alias | cancer/testis antigen 5.2; cancer/testis antigen family 5, member 2a; CT5.2; CT5.2A; CT52; HD21; HOM-MEL-40; LOC103690782; MGC119055; MGC15364; MGC3884; Protein SSX2; putative protein SSX10; sarcoma, synovial, X-chromosome-related 2; SSX; SSX family member 2; SSX2; SSX2A; SSX2B; Synovial sarcoma, X breakpoint 2; synovial sarcoma, X breakpoint 2B; tumor antigen HOM-MEL-40 |
| Gensymboler | Ssx2 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?
