missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSX5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12401-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The product of this gene belongs to the family of highly homologous synovial sarcoma X (SSX) breakpoint proteins. These proteins may function as transcriptional repressors. They are also capable of eliciting spontaneously humoral and cellular immune responses in cancer patients, and are potentially useful targets in cancer vaccine-based immunotherapy. SSX1, SSX2 and SSX4 genes have been involved in the t(X;18) translocation characteristically found in all synovial sarcomas. This gene appears not to be involved in this type of chromosome translocation. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
SSX5 | |
Polyclonal | |
Unconjugated | |
SSX5 | |
Protein SSX5, SSX5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6758 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60225 | |
SSX5 | |
SSX5 Fusion Protein Ag3095 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |