missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ SSYA10-001

Description
SSYA10-001 is a potent coronavirus helicase inhibitor (IC50 values are 0.046 μM and >3 μM for unwinding and ATPase, respectively). Inhibits SARS-CoV-2 Nsp13 helicase (IC50 = 3.5 μM), blocks dsDNA and dsRNA unwinding activities of Nsp13 (IC50 values are 5.3 μM and 5.7 μM respectively). In a SARS-CoV replicon assay SSYA10-001 inhibits (+) sense but not (-) sense RNA synthesis (EC50 = 8.95 μM). Also inhibits human papillomavirus E6 (IC50 = 10 μM)
Specifications
Specifications
| Kemiskt namn eller material | 2,4-Dihydro-5-[[(2-nitrophenyl)thio]methyl]-4-(2-propen-1-yl)-3 H-1,2,4-triazole-3-thione |
| CAS | 675104-49-1 |
| Kvantitet | 50 mg |
| Mål | Antiviral Compounds |
| Molekylformel | C12H12N4O2S2 |
| Renhet | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?