Läs mer
Tocris Bioscience™ st-Ht31
GREENER_CHOICE
Inhibits PKA/AKAP interactions; cell permeable
Brand:  Tocris Bioscience™ 6286/1
Encompass
Produktkod. 15746389
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
st-Ht31 is a stearated form of the peptide Ht-31. Inhibits the interaction between the RII subunits of cAMP-dependent PKA and A-kinase anchoring protein (AKAP) in cell extracts. Cell permeable.
Specifikationer
Specifikationer
188425-80-1
Store at -20°C
2798.27
1 mg
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer