Learn More
Thermo Scientific™ ST16R TX-400 Cell Culture Centrifuge Package
Exceptional capacity, ergonomic features and value.
Brand:  Thermo Scientific™ 50126391
Additional Details : Weight : 0.80000kg
Product Code. 11824243
  • 102504.92 SEK / Each
Remote Stock
(10 days availability): 8
Add to basket
Includes: Sorvall ST 16R, 230V (Mfr PN: 75004380); TX-400 rotor, Slide coat (Mfr PN: 75003181); 400mL Round buckets (set/4) - for TX-400 rotor (Mfr PN: 75003655); Sealing caps for TX-400 bucket (set/4, includes grease) (Mfr PN: 75003656); Adapters 4 x 50 conical, set of 4 (Mfr PN: 75003683); Adapters 9 x 15mL conical, set of 4 (Mfr PN: 75003682)
Description
Description
The Thermo Scientific Sorvall ST 16 general purpose centrifuge series is perfect for everyday sample processing, including clinical protocols, cell culture applications and microplate processing.
Specifications
Specifications
Sorvall ST 16R, 230V (Mfr PN: 75004380); TX-400 rotor, Slide coat (Mfr PN: 75003181); 400mL Round buckets (set/4) - for TX-400 rotor (Mfr PN: 75003655); Sealing caps for TX-400 bucket (set/4, includes grease) (Mfr PN: 75003656); Adapters 4 x 50 conical, set of 4 (Mfr PN: 75003683); Adapters 9 x 15mL conical, set of 4 (Mfr PN: 75003682)
ST16R TX-400 Cell Culture Centrifuge Package
Videos