missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ST6GALNAC6 Recombinant Protein

Brand:  Abnova™ H00030815-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179322

  • 4515.00 SEK / 10µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH06564
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
62.37
8
Glutathione Sepharose 4 Fast Flow
10 ug
MACSRPPSQCEPTSLPPGPPAGRRHLPLSRRRREMSSNKEQRSAVFVILFALITILILYSSNSANEVFHYGSLRGRSRRPVNLKKWSITDGYVPILGNKTLPSRCHQCVIVSSSSHLLGTKLGPEIERAECTIRMNDAPTTGYSADVGNKTTYRVVAHSSVFRVLRRPQEFVNRTPETVFIFWGPPSKMQKPQGSLVRVIQRAGLVFPNMEAYAVSPGRMRQFDDLFRGETGKDREKSHSWLSTGWFTMVIAVELCDHVHVYGMVPPNYCSQRPRLQRMPYHYYEPKGPDECVTYIQNEHSRKGNHHRFITEKRVFSSWAQLYGITFSHPSWT
RP11-203J24.3/SIAT7F/ST6GALNACVI
ST6GALNAC6
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
30815
ST6GALNAC6 (Human) Recombinant Protein (P02)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ST6GALNAC6
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers