Learn More
Thermo Scientific™ Stability Chamber, 311 L, Stainless Steel 1.4301, 2B
Temperature Range: 0°C to 60°C; Relative Humidity (RH): <95% at 37°C; Light: No Lighting
Brand:  Thermo Scientific™ 3907
Additional Details : Weight : 260.79999kg
Product Code. 15480430
  • 235165.00 SEK / Each
Estimated Shipment
21-09-2020
Add to basket
Description
Description
  • Optimum uniformity and a wide temperature range designed for easy compliance with regulatory requirements.
  • Rugged construction and durable shelving for long life.
  • Broad range of options to create the perfect fit for your applications.
Specifications
Specifications
No Lighting
15.9 kg (slide in/out); 22.7 kg (stationary)
CE
311 L
0°C to 60°C
Powder Coated Cold Rolled Steel
230 V, 50/60 Hz; 15.0 A
Digital, dual LED (direct read in % RH)
Gravity to 2.76 bar (40 psi)
65.6 cm
CEE 7/7
Non-CFC, R134A refrigerant
324.3 kg
RTD
230 V
260.8 kg
Stability Chamber
Audible/visual
15.0 A
Stainless Steel, Type 1.4301
<95% at 37°C
Digital Dual LED
Microprocessor, PID
Bulk polymer
Three shelves
3/11
Fill Port: 1⁄8 in. female NPT; Drain Port (bottom): 3⁄8 in. FPT & 3⁄8 in. O.D. P-trap right rear
0.5 sq. m
±0.1°C @ 25°C to 37°C (77°F to 98.6°F)
±0.3°C @ 25°C to 37°C (77°F to 98.6°F)
3.78 L/hour (1 gallon/hour)
77.8 cm