missing translation for 'onlineSavingsMsg'
Learn More

Stachyose hydrate, technical, ACROS Organics™

Brand:  Acros Organics 453140010

Additional Details : CAS Number : 54261-98-2 Weight : 0.01000kg

Quantity 1g

 View more versions of this product

Product Code. 15229836

  • 1632.56 SEK / 1g


Estimated Shipment
09-11-2020
Add to basket

Specifications

Chemical Identifiers

54261-98-2
666.58
131801001
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)OC4(C(C(C(O4)CO)O)O)CO)O)O)O)O)O)O)O)O)O)O.O.O.O
C24H42O21·xH2O
XQDOUZBIGFWLCL-UHFFFAOYSA-N
2-[[6-[[6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;trihydrate

Specifications

Stachyose hydrate
+ 129.00 (20.00°C c=4, H2O)
12% max.
1g
C24H42O21·xH2O
XQDOUZBIGFWLCL-UHFFFAOYSA-N
2-[[6-[[6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;trihydrate
131801001
Technical
Authentic
+ 129.00
95.0°C to 105.0°C
54261-98-2
Solubility in water: soluble.
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)OC4(C(C(C(O4)CO)O)O)CO)O)O)O)O)O)O)O)O)O)O.O.O.O
666.58
666.58
Videos
Documents