Learn More
Ohaus™ Stackable Microplate Holder
OHAUS™ Stackable Microplate Holder allows you to work with up to four microplates at one time.
Brand: Ohaus™ 30400212
Additional Details : Weight : 0.50000kg
Description
- For use with vortex mixers
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up four microplates
Specifications
4 Microplates | |
Plasitc | |
4 |
Microplate Holder | |
4 Place Stackable Microplate Holder |