missing translation for 'onlineSavingsMsg'
Läs mer

Stainless Steel, Alloy 253 MA , S30815, ARMI∣MBH

Produktkod. 18033970
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18033970 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18033970 Leverantör ARMI/MBH Leverantörsnummer IARM316AG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Stainless Steel, Alloy 253 MA , S30815, Ø38x19mm
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Zinc/Calcium/Oxygen/Aluminium/Tin/Arsenic/Tungsten/Phosphorus/Vanadium/Cerium/Carbon/Cobalt/Nitrogen/Copper/Molybdenum/Manganese/Silicon/Nickel/Chromium/Lead/Boron/Titanium/Niobium/Tantalum
CAS 7439-89-6
Kvantitet 1 Ea.
Form Disc
Koncentration eller sammansättning (efter analyt eller komponenter) Zinc 0.0008 %/Calcium 0.0017 %/Oxygen 0.0052 %/Aluminium 0.006 %/Tin 0.006 %/Arsenic 0.007 %/Tungsten 0.022 %/Phosphorus 0.023 %/Vanadium 0.042 %/Cerium 0.064 %/Carbon 0.07 %/Cobalt 0.118 %/Nitrogen 0.16 %/Copper 0.19 %/Molybdenum 0.25 %/Manganese 0.61 %/Silicon 1.5 %/Nickel 10.81 %/Chromium 21.07 %/Lead (0.0001) %/Boron (0.0003) %/Titanium (0.002) %/Niobium (0.003) %/Tantalum (0.003) %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.